Fine Diamond Scriber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62108-ST 62108-BT 62107-BT
100502-476EA 128.12 CAD
100502-476 100497-840 100497-836
Fine Diamond Scriber, Electron Microscopy Sciences
Pens Stylus and Scribers
This diamond scriber is the ultimate scribing instrument offering the most precision for the most delicate applications.
  • Fine scribing under the microscope - for silicon wafers and glass coverslips
  • Precision scribing and repairing - for thin film circuits and microcircuits, and more
It is available in straight or bent configurations.The integrated 6.35 mm anodized hexagonal (non-rolling) aluminum handle has a positive finger grip in the form of grooves for easy control and nonslippage. Each diamond scriber is packaged in a protective plastic tube.