Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62118-12 62118-02
100490-142EA 62.32 CAD
100490-142 100497-846
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Abrasives
A plastic handle, 267 mm (10 1/2") long, holds strips of emery paper or cloth 279 x 25.4 mm (11 x 1"). Strips are easily replaced. Comes complete with 2 strips/set (grit 120 and 240).