Fairs NG5 Graticule, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68033-19 68033-21 68033-16
100499-280EA 686.19 CAD
100499-280 100499-282 100499-278
Fairs NG5 Graticule, Electron Microscopy Sciences
Graticules, Reticles and Grids
Designed to extend the sizing range of globe and circle graticules. Example: Used in conjunction with NG2 the overall size range =128:1. The circles increase by 2.
Caution: Note: Both graticules would have to be used with the same microscope, eyepiece and objective.