Kellner and Huygenian Focusing Eyepieces, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67990-F12 67990-F7 67990-F10
100499-058EA 880.6 CAD
100499-058 100499-060 100499-056
Kellner and Huygenian Focusing Eyepieces, Electron Microscopy Sciences
Microscope Eyepieces
Focusing eyepieces for microscopes.
Huygenian Eyepiece×7, composed of two plano convex lenses, separated by an air space. The image plane is between the two lenses. The top lens is adjusted to bring the graticule into focus.
Kellner Eyepiece×10, the F10 eyepiece, which fits most standard monocular microscopes, is specially designed to receive eyepiece graticules of 16 mm diameter. This allows the graticule to be brought into sharp focus when superimposed on the object image, which is often difficult to achieve with fixed focus eyepieces. The graticule is held by a screw-on retaining ring and is easily removed for cleaning or replacement for another pattern. Composed of a convex lens and an achromatic lens. The image plane is below the lens elements. The use of an achromatic lens provides a larger field of view, better color correction and substantial eye relief. The top lens adjusts to focus on the graticule.Graticule: 16 mm diameter
Kellner Type Eyepiece (Wide Field×10), F12 with high eye point (useful for spectacle wearers). Non -focusing wide field.Graticule:19 mm diameter.