EMS Lucite® Film Racks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74891-24 74891-25 74891-18 74890-18 74895-12
100494-206EA 177.34 CAD
100494-206 100494-208 100494-204 100494-202 100494-210
EMS Lucite® Film Racks, Electron Microscopy Sciences
Racks Film Racks
Fabricated from genuine Lucite®; these racks fit into our 5×7" developing tanks. One end of the rack is extended in length with a cut-out hole to serve as a handle.
The racks are designed to accommodate a number of sheets per rack without changing the length of the rack. As the number of the films per rack increases, the space between them decreases.