SB50 Grid Storage Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71136-12 71135-12 71136-01 71135-01
100489-236EA 665.84 CAD
100489-236 100489-234 100493-132 100493-130
SB50 Grid Storage Box, Electron Microscopy Sciences
Boxes Storage Boxes
The clear cover can be rotated smoothly through 360 degrees once the slight initial resistance of the park position has been overcome exposing a maximum of 2 or 3 diamond shaped slots at any one time
The 50 diamond shaped slots have an alphanumeric referencing system. Each box is supplied with an index record card for additional information. The storage boxes are designed to be stacked, the base locating precisely over the face of another box. The material the bases are made from have been chosen due to their anti-static properties. The clear cover has self-lubricating properties.