Embedding Rings, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62350-Y 62350-G 62350-W 62350-P
100500-980BX 105.92 CAD
100500-980 100500-974 100500-978 100500-976
Embedding Rings, Electron Microscopy Sciences
Embedding Cassettes Embedding Rings
Molded from the highest quality plastic, our Embedding Rings fit most base molds. For added convenience, there are four colors from which to choose Blue, Pink, Green, Yellow. The large writing surface further eases identification and handling. The base measures 1 5/8 x 1 1/8" and the ring measures 25 x 25 mm (1 x 1").