Accessories for BEEM™ Capsules; Embedding, Size 00, 8 mm I.D., Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69915-06 69916-06 69916-01 70022-06 70022-12 69915-01 70022-01 69915-12 70022-04 69917-01-3 69917-06-3 69917-12-3
100490-714EA 226.27 CAD
100490-714 100490-718 100492-864 100488-952 100488-954 100492-862 100492-884 100490-716 100488-950 103301-920 103302-262 103305-110
Accessories for BEEM™ Capsules; Embedding, Size 00, 8 mm I.D., Electron Microscopy Sciences
Embedding Cassettes Tissue Capsules
Fabricated form polypropylene, a direct substitution for the BEEM® capsule holder
15 positions. Fits embedding capsule size ʽ00’ (8.5 mm diameter).
Order Now
SPECIFICATIONS
Environmentally Preferable