Embedding Blocks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62363-02 62363-03 62363-01 62363-04
100498-056EA 179.11 CAD
100498-056 100498-058 100498-054 100498-060
Embedding Blocks, Electron Microscopy Sciences
Embedding Cassettes Embedding Rings
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.