DuraSeal™ Polyethylene Stretch Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70994-20 70994-04 70994-05 70994-01 70994-02 70994-06 70994-14
100493-038EA 33.13 CAD
100493-038 100488-114 100501-794 102097-226 102097-228 102097-230 103303-504
DuraSeal™ Polyethylene Stretch Film, Electron Microscopy Sciences
Seals General Purpose Sealing Films
This Durable heat-resistant, solvent-proof laboratory stretch film forms an airtight heat-resistant barrier.
  • Microwave compatible and tear resistant
  • Chemically resistant to all solvents and caustic agents
  • Withstands a temperature of 100 °C
  • Able to stretch 500 to 800% before breaking or tearing
  • Able to cling to self following stretching