Gilder Double Grids (Oyster), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
GD1020-NI GD50-CU GD1010-NI GD1010-CU
100495-728EA 164.81 CAD
100495-728 100495-734 100495-724 100495-722
Gilder Double Grids (Oyster), Electron Microscopy Sciences
Graticules, Reticles and Grids
Double grids are used mostly in metallurgical applications for supporting thin metal foils. These grids have a curved securing tab which folds to the curvature of the 'sandwiched' grid.
  • Material: Copper (Cu) or Nickel (Ni)
  • Measures 3.05 mm in diameter
  • Four configurations are available