Double Clamp, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62123-02 62123-01
100497-854EA 47.18 CAD
100497-854 100497-852
Double Clamp, Electron Microscopy Sciences
Clamps General Purpose Clamps
A versatile tool which can be adjusted to any position and firmly hold small items of any shape.
Two adjustable arms with alligator jaws in a heavy base. Double Clamp is also available with a 2 ½" diameter magnifier attached.