Disposable Microtome Blades; Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63061-01 63063-10 63063-01 63062-01 63061-10 63062-10 63064-10 63064-01
100500-984BX 486.48 CAD
100500-984 100498-336 100500-988 100500-986 100498-332 100498-334 100498-338 100500-990
Disposable Microtome Blades; Stainless Steel, Electron Microscopy Sciences
Razors and Blades
Disposable microtome blades are coated with PTFE and represent a new standard in sectioning quality.
Blades are available in three different types.
Standard Blade: For routine tissue samples, the standard blade offers an unsurpassed level of quality at an affordable price.
Premium Blade: When more difficult specimens are encountered, simply switch to the premium type-a heavy duty blade which allows to achieve ribbons of unsurpassed quality.
The Edge: Years of research went into choosing the optimum stainless steel and edge combination. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine.