Dip Miser, For Slide Coating, 10 ml Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70520
70530
70510
100491-620EA
183.63
CAD
100491-620
100499-696
100491-618
Dip Miser, For Slide Coating, 10 ml Capacity, Electron Microscopy Sciences
Stains
Microscopy and staining reagents
Slide coating cup with stand and cleaning brush. Glass cup is Pyrex and is designed to conserve expensive autoradiographic emulsion when coating glass slides. Needs only 1/4 the volume of solution previously required. Single piece holder permits immersion in a water bath and provides a stable darkroom stand to prevent spills, and faster temperature adjustment of the emulsion.