PrecISIon Diamond Wafering Saw Blades, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50266-CL7 50266-MH3 50266-MH4 50267-CH4 50265-CH6 50265-CL5 50265-CL4 50265-FH3 50266-FH4 50267-FH4 50265-CH7 50265-ML3
101411-830EA 1432.97 CAD
101411-830 101411-834 101411-836 101411-838 101411-806 101411-810 101411-808 101411-814 101411-832 101411-840 102092-782 102092-784
PrecISIon Diamond Wafering Saw Blades, Electron Microscopy Sciences
Razors and Blades
These bonded blades are constructed of an inner metal core and an outer rim, this rim is bonded with diamond particles
High Concentration (H/C) blades are for general laboratory use, either low or high saw’s speed while low concentration (L/C) blades are for sectioning hard and brittle materials, such as ceramics, silicon, glass, minerals.
These blades come in high and low concentrations of diamond particles to handle various sectioning requirements.