Steam Autoclaves Imprinted Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77017-10 77016-10
100488-168EA 85.23 CAD
100488-168 100488-174 100488-170
Steam Autoclaves Imprinted Tape, Electron Microscopy Sciences
Labels
Indicate the status of all laboratory items to be sterilized with these tape labels.
The word 'STERILIZED' appears after processing at 250 °F under normal sterilization conditions. Has 3" core.