Diamond Dissecting Knives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72027 72025 72026 72029 72032 72030 72031
100491-900EA 3465.34 CAD
100491-900 100491-896 100491-898 100491-904 100491-910 100491-906 100491-908
Diamond Dissecting Knives, Electron Microscopy Sciences
Knives
DIATOME has now begun to manufacture diamond knives and blades for microsurgery, dissection, and its related applications.
  • Ultra-sharp blade cuts with minimum pressure, without tearing or damaging the specimen and reduces the risk of specimen distortion
  • Unique pressure mechanism allows blade actuation with one hand
  • The handle is made of titanium and is coated by high vacuum with titanium nitride to harden the surface.
  • The knife is corrosion free and is completely resistant to saline solutions.
  • It can be sterilized in an autoclave up to 200°C.
  • Special tray for sterilization is available
Available in these ten following styles: angled (.50 mm thickness), mini-angled (.20 mm thickness), lancet (.50 mm thickness), mini-lancet (.20 mm thickness), double-lancet (.50 mm thickness), mini-double-lancet (.20 mm thickness), tri-facet (.50 mm thickness), mini-tri-facet (.20 mm thickness), round (.30 mm thickness) and half-round (.20 mm thickness) styles.
The knives are of the same quality that DIATOME customers have become accustomed. Distinctive designs combined with DIATOME'S expertise in diamond technology and manufacturing assures you the best quality knife and being able to use it for many years.
If your diamond is damaged or broken, we can resharpen it or replace it and guarantee that the repaired knife will be of the same standards of perfection as the original.