Cotton Tipped Applicators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72310-10 72310 72307 72310-S
100499-648EA 63.94 CAD
100499-648 100489-878 100496-340 100493-712
Cotton Tipped Applicators, Electron Microscopy Sciences
Swabs Swabs for Clinical Use
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.