Coverglass Staining Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72240
100491-942EA 505.41 CAD
100491-942
Coverglass Staining Rack, Electron Microscopy Sciences
Staining Boxes
Coverglass staining rack is made from polished stainless steel.
  • Holds five of 22×24 mm coverglasses
  • Comes with a handle, 12.7 mm (5") long
This rack is specially designed to be used with coplin jars.