Cloth; Lint Free, Twill Jean, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71750 71753 71752
100488-444EA 126.29 CAD
100488-444 100488-448 100488-446
Cloth; Lint Free, Twill Jean, Electron Microscopy Sciences
Wipes Reusable Cleaning Cloths Cloths for General Applications
A twill jean cotton wipe made from 100% cotton fibre that is woven in an extremely tight construction.
  • Highly absorbent
  • Thermal stability, good for high heat applications
  • Excellent wipe ability
  • Acid resistant and solvent compatible
  • Low metallic ion content
  • Low static generation
The extractable trace elements are effectively reduced in the cleaning process which removes impurities in the cotton as well as any starches or binders used in the manufacturing process.