Carbon Fiber and Cord, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
91045 91046 91045-100 91046-1 91046-100 91047-1
100492-488EA 69.14 CAD
100492-488 100492-490 102099-906 102099-908 102099-912 102099-914
Carbon Fiber and Cord, Electron Microscopy Sciences
Cables
Carbon Fibre Cord - high purity: Used With: Most carbon coaters where thick carbon cord is used.
Sometimes referred to as 'braid’, carbon fibre consists of eight separate strands of individual carbon fibre braided together. The fibre is manufactured so that, when cut, the ends resist unravelling. The fibre density is about 1.6 g/m.
Carbon Fibre Cord - high purity: Used With: Most carbon coaters where thick carbon cord is used.
Carbon Fibre Cord- standard grade: Convenient to use, giving thick coating using a single length. Measures 1 m length.
Carbon Fibre - fine strands: Convenient to use, giving thin to medium coating using a single strand or several lengths. Measures 1 m length.