Capsules, Gelatin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70101 70105 70104 70100 70102 70103 70110
100488-358EA 204.61 CAD
100488-358 100488-366 100488-364 100488-356 100488-360 100488-362 100488-368
Capsules, Gelatin, Electron Microscopy Sciences
Moulds Moulds for Histology
Natural, hard gelatin capsules are used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. The gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36 to 38 °C, within 2 minutes in water at 37 °C or in artificial gastric juice.