BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70000-B 70010-B 70020-B
102096-550EA 21.08 CAD
102096-550 102096-556 102096-558
BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
Embedding Cassettes Tissue Capsules
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.