Calibrated Loops, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62430-20 62430-30 62430-10
100498-078EA 1060.46 CAD
100498-078 100498-080 100498-076
Calibrated Loops, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
The 0.01 mL loops are calibrated to A.P.H.A. Standards and are made from 19 gauge B and S platinum wire with 5% rhodium. The 0.001 mL loops is made from 26 gauge B and S platinum with 15% iridium.