Burnishers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77615-01 77615-02 77615-03
100494-388EA 34.55 CAD
100494-388 100494-390 100494-392
Burnishers, Electron Microscopy Sciences
Polishers and Tumblers
These burnishers are especially designed for dry transfer products. They are pressure adjustable allowing you to set and maintain the proper pressure for any burnishing style.
For maximum adhesion, place the backing sheet over the newly transferred letters and reburnish with the "bone end." The rolling tip burnisher is ideal for large transfers, the standard tip is ideal for standard work, and the fine tip for detail work.