Brushes, Red Sable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66105 66100-02 66100-01 66100-50 66100-00 66100-30 66100-03 66100-40
100496-236EA 65.89 CAD
100496-236 100498-902 100498-900 100502-550 100498-898 100498-908 100498-904 102095-926
Brushes, Red Sable, Electron Microscopy Sciences
Brushes
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.