Broken Crosslines, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67985-16 67985-19 67985-21
100499-026EA 280.92 CAD
100499-026 100499-028 100499-030
Broken Crosslines, Electron Microscopy Sciences
Graticules, Reticles and Grids
Broken lines enable fine detail to be seen at the breaks. A thin boundary would be lost behind a continuous line.