Breakable Double Edge Razor Blades, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72004
100500-640BX 176.04 CAD
100500-640
Breakable Double Edge Razor Blades, Electron Microscopy Sciences
Razors and Blades
Breakable double edge razor blades made from carbon steel.
The blades break easily and cleanly. Rectangular shape, square ends.
Measures L x W x T: 43 x 22.2 x 0.10 mm.
Packaging: Packaged 100 blades in a box.