MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems
A set of 18 standards, a Faraday Cup included. Mounted in 25 or 32 mm diameter brass block. Includes Al, BaF2, BaSO4, Bi, BN, C , CaCO3, CaSO4, FeS2, InP, KBr, KCl, MgO, NaCl, Se, SiO2, Ti, V.
All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.