Beryllium, substrate planchets
Supplier: Electron Microscopy Sciences
|
Danger
|
76010
76016
76017
100492-280EA
577.73
CAD
100492-280
100492-286
100492-288
Beryllium, substrate planchets
Beryllium
These planchets are prepared from high purity beryllium (min. 98.5%) by electro-fusion to provide vacuum tight (~1x10-9atm-cm3/sec) and ±10% dimensional tolerances.
Formula:
Be Boiling Pt: 2970 °C (5378 °F) Melting Pt: 1278 °C (2332 °F) Density: 1.85 g/cm³ (15.438 lbs/gal.) |
MDL Number:
MFCD00134032 CAS Number: 7440-41-7 UN: 1567 ADR: 6.1,II |
Specification Test Results
Beryllium Assay |
|
Beryllium Oxide | 1.50 % max |
Iron Content | 0.1300 % max |
Aluminum Content | 0.10 % max |
Magnesium Content | 0.0800 % max |
Silicon Content | 0.0600 % max |
Carbon Content | 0.15 % max |
Other Metallic– each | 0.0400 % max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...