Benchtop Mat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64446 64445
100500-862BX 813.23 CAD
100500-862 100502-686
Benchtop Mat, Electron Microscopy Sciences
Surface Protectors Bench Protectors
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions.
  • Sheets can be rolled up and discarded after use
  • Ideal for daily use in EM labs
Benchtop mat comes in a roll 50.8 cm (20") wide and 15.2 m (50') long in a dispenser carton, or a cut size of 20×24" sheets.