BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69910-01 69910-05 69910-10
102096-532EA 21.31 CAD
102096-532 102096-534 102096-536
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Embedding Cassettes Tissue Capsules
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Order Now
SPECIFICATIONS
Environmentally Preferable