Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60952 60960 60950 60962
100503-198EA 214.99 CAD
100503-198 100503-278 100503-196 100503-200
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Beakers
Made from heavy gauge polypropylene
  • Resistant to most chemicals
  • Frosted marking area
  • Customary and metric graduations
  • Anti-drip ridge
Suitable for parafilm cover.
Order Now
SPECIFICATIONS
Environmentally Preferable