All Glass Nebulizer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70505-05 70506-05 70506-01 70505-01
100499-568EA 20524.25 CAD
100499-568 100489-184 100492-968 100492-966
All Glass Nebulizer, Electron Microscopy Sciences
Atomizers
An all glass unit for the simple production of microdroplets
An object is held vertically in front of the nebulizer outlet and by squeezing the atomizer a fine spray is created. The nebulizer set comes with an All Glass Nebulizer bulb and Atomizer.