ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50425-10 50425-25
100490-030EA 607.81 CAD
100501-976 100490-030
ACLAR®-Fluoropolymer Films, Electron Microscopy Sciences
Film Processing Supplies
ACLAR® is made from fluorinated-chlorinated resins that provide an exceptional moisture barrier. ACLAR® is crystal clear, biochemically inert, highly resistant to most chemicals and sterilizable by heat or radiation. ACLAR® is used widely in pharmaceutical, medical, sensitive electronics and military packaging.
  • Crystal clear, high UV transparency- ideal for use in UV curing of embedding resin in microscopy
  • Chemically stable and biochemically inert- the product of choice for growing cell cultures
  • Low dielectric constant, high electric strength and dissipation factor– offers excellent cell attachment even through lengthy processing procedures
  • Low surface energy–and separates easily from epoxy
  • Exhibits no detectable autofluorescence– ideal for fluorescent microscopy
  • A non-flammable, non-aging co-polymer– which is suited for sterilization by heat or UV
  • Crystalline melting point is 206 °C (403 °F)– stable in the SEM
  • Flexible and soft– can be sectioned without damage to the ultramicrotome knives
There are two film types–the homopolymer ACLAR® Rx Series and the copolymers ACLAR® 33C.
ACLAR® 33C is a copolymer film consisting primarily of chlorotrifluoroethylene (CTFE). It offers an outstanding moisture barrier, excellent chemical resistance and minimal dimensional change (±2%), making it the best choice for use in microscopy.
Aclar® UltRx 2000 is a 2.00 mil (51 micron) PCTFE homopolymer, high performance barrier film for the pharmaceutical and medical markets. It thermoforms well on conventional blister packaging equipment and provides the best barrier of any clear film.