Ampoule Cracker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60605-20 60605-10 60605-04
100490-064EA 267.37 CAD
100490-064 100502-770 100490-062
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Openers
The Ampoule Cracker™ is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules.
Simply slip the Ampoule Cracker™ over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.