Permanox Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70356 70350 70353
100499-892CS 3689.33 CAD
100499-892 100488-378 100488-380
Permanox Dish, Electron Microscopy Sciences
Culture Dishes Petri Dishes
Permanox is a strong, biologically inert material, resistant to nonchlorinated hydrocarbons, acetone, and many other EM reagents. Our EMbed-812 and other embedding resins can be used to embed cells in these dishes with excellent results. Permanox has not been shown to be autofluorescent, and has a low oxygen solubility which makes it suitable for experiments performed under anoxic conditions. The dishes have high optical quality that provide excellent and consistent viewing where microscopic examination is required.