Centrifuge Screw Cap Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64760-10 64763-01 64760-01 64762-01 64762-20 64764-01 64763-02
100501-146BG 32.55 CAD
100501-146 100502-022 100498-864 100501-148 100501-150 100501-156 100502-024 100501-152 100501-154
Centrifuge Screw Cap Tubes, Electron Microscopy Sciences
Tubes Centrifuge Tubes
Premium quality general purpose centrifuge tubes with printed graduations.
Available in two different volumes:
50 ml: Made from premium polypropylene copolymer for excellent chemical resistance; withstands temperature extremes of –40 to 125 °C; high centrifugation of 6000RCF with exceptional clarity.
15 ml: Available in poly-styrene or polypropylene. Polystyrene offers glass-like clarity and can withstand 1800RCF. Polypropylene has excellent chemical resistance and can withstand 6400 RCF.
These tubes are available in bulk, bag pack, or in expanded styrene foam trays. As well they areMore Product Information