Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71539-01 71539-10 71541 71540-CS 71541-CS
100489-334EA 76.71 CAD
100489-334 100489-336 102091-220 102097-600 102097-602
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Slide Mailers
The slide mailers are available in sizes to hold one, two, or four 3 x 1" glass slides.
1-Slide Solid Cardboard Slide Mailer is made from 0.050” (1.29 mm) solid cardboard, measures 33/4 x 15/8” when folded. The mailer holds one 3 x 1” glass slide with or without cover glass and has a thumb groove for easy removal. A special cut corner identifies the top of the mailer.
2-Slide Solid Cardboard Slide Mailer is made from .085” (2.17 mm) cardboard and measures 3 x 41/4” when folded. This mailer is unprinted. Upon request the mailer can be printed with warnings for medical specimens. Each mailer conforms to U.S. Postal regulations regarding minimum size for First Class.
4-Slide Solid Cardboard Slide Mailer has the same construction as 2-slide mailer above but holds 4 of 3 x 1" slide.