Dumont Tweezers Style SS135, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0304-SS135-CO 0304-SS135-PO
102091-722EA 64.39 CAD
102091-722 102091-724
Dumont Tweezers Style SS135, Electron Microscopy Sciences
Forceps
Long narrow shanks, thin tips. Length: 135 mm.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.