Dumont Tweezers Style 0, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0302-0-PO 0304-0-CO
102091-622EA 81.97 CAD
102091-622 102091-664
Dumont Tweezers Style 0, Electron Microscopy Sciences
Forceps
Antimagnetic is composed of C, Cr, Ni and Mo
Geneva pattern, thin tips. Length: 115 mm.
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.