Dumont Tweezers Style N5AC, Dumont Switzerland, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72856-D 72856-DZ 0202-N5AC-PO 0207-N5ACTI-PO
100488-320EA 162.25 CAD
100488-320 100499-820 102091-570 76375-934
Dumont Tweezers Style N5AC, Dumont Switzerland, Electron Microscopy Sciences
Forceps
Anti-capillary, type 5, super thin tips.
  • Hand made of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. Lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.
Dumoxel is composed of C, Cr, Ni, Mo and Cu. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion.More Product Information