Dumont Tweezers Style N5, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72854-D 72854-DZ 0103-N5-PO 0202-N5-PO
100501-168EA 120.35 CAD
100501-168 100499-818 102091-548 102091-572
Dumont Tweezers Style N5, Electron Microscopy Sciences
Forceps
Type 5 is 108mm, super thin tips.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400° C (DIN 50 914) and is suitable for autoclave sterilization at 270°C. Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. Lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.