Tweezers Style 7, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72803-12 72800-DZ 72800-D 72803-01 0208-7-PO
100499-810DZ 1704.05 CAD
100499-810 100499-798 100488-305 100493-840 102091-612
Tweezers Style 7, Electron Microscopy Sciences
Forceps
Dumont positive action Style 7 tweezers have thin curved tips.
  • Hand made of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Dumoxel® is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel® also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to allMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable