Dumont Tweezers Style 55, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72707-01 72707-12 0203-55-PO
100493-838EA 155.85 CAD
100493-838 100499-782 102091-580
Dumont Tweezers Style 55, Electron Microscopy Sciences
Forceps
This Dumont patented alloy is composed of C, Cr, Mo, Mn, Co, Ni and Si
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400° C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
This Dumont patented alloy is composed of C, Cr, Mo, Mn, Co, Ni and Si. Dumostar is more elastic and more resistant to corrosion than the best stainless steels. It is also perfectly compatible with human tissues and is resistant to mineral and organic acids as well to salt. Although slightly more expensive than other alloys, Dumostar is 100% antimagnetic and is resistant to sterilization temperatures of up to 500°C.This is the most cost effective and appropriate alloy for laboratory use.
Dumostar is more elastic and more resistant to corrosion than the best stainless steels. It is also perfectly compatible with human tissues and is resistant to mineral and organic acids as well to salt. Although slightly more expensive than other alloys, Dumostar is 100% antimagnetic and is resistant to sterilization temperatures of up to 500°C.This is the most cost effective and appropriate alloy for laboratory use.