Dumont Tweezers Style 24, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72880-DS 72880-DB
100493-860EA 154.29 CAD
100493-860 100493-858
Dumont Tweezers Style 24, Electron Microscopy Sciences
Forceps
Strong tips with inside serrations. Length: 150 mm.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400°C (DIN 50914) and is suitable for autoclave sterilization at 180°C.