Tweezers Style 4, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72695-DZ 72695-D 0209-4-PO 0508-4-PO
100499-756DZ 1073.74 CAD
100499-756 100501-804 102091-618 102091-746
Tweezers Style 4, Electron Microscopy Sciences
Forceps
Dumont positive action Style 4 tweezers have very thin tips.
  • Handmade of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Dumoxel® is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel® also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel® is 95% antimagnetic, resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.
Dumostar®, Dumont's patented alloy, is composed of C, Cr, Mo, Mn, Co, Ni and Si. Dumostar is more elastic and more resistant to corrosion than the best stainless steels. It is also perfectly compatible with human tissues and is resistant to mineral and organic acids as well to salt. Although slightly more expensive than other alloys, Dumostar is 100% antimagnetic and is resistant to sterilization temperatures of up to 500 °C. This is the most cost effective and appropriate alloy for laboratory use.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400 °C (DIN 50914) and is suitable for autoclave sterilization at 180 °C.