Dumont Tweezers Style 3C, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72681-D 72680-D 72683-12 72680-DZ
100488-262EA 85.65 CAD
100488-262 100488-258 100499-738 100499-726
Dumont Tweezers Style 3C, Electron Microscopy Sciences
Forceps
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400°C (DIN 50914) and is suitable for autoclave sterilization at 180°C.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented byMore Product Information