Dumont Tweezers Style N4, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0103-N4-PO
102091-546EA 117.05 CAD
102091-546
Dumont Tweezers Style N4, Electron Microscopy Sciences
Forceps
Type 4, very thin tips, 108 mm.
Dumoxel is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400° C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.