Tweezers Style 2, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0103-2-PO 0109-2-PO
102091-524EA 86.11 CAD
102091-524 102091-554
Tweezers Style 2, Electron Microscopy Sciences
Forceps
Dumont style 2 positive action tweezers have strong long tips.
  • Handmade of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Dumoxel® is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel® also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel® is 95% antimagnetic, resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.